Learn More
SI Analytics™ Measuring And Storing Vessel
Product Code. 10326674 Shop All SI Analytics Products
Change view
Click to view available options
For Use With (Equipment):
Electrodes with 12 mm dia.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10326674 Electrodes with 12 mm dia. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10326674 Supplier SI Analytics™ Supplier No. 285123170
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
SI Analytics™ Measuring And Storing Vessel is a plastic receptacle with squeezing ring sealing and bayonet fastener for electrodes with a 12mm diameter.
  • Plastic receptacle
  • includes squeezing ring sealing and bayonet fastener
  • Suitable for electrodes with 12mm diameter
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Electrodes with 12 mm dia.
Type Measuring and Storing Vessel
Includes Squeezing Ring, Sealing and Bayonet fastener