Learn More
ME2 Rabbit anti-Human, Polyclonal, Proteintech
Product Code. 16883465 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16883465 20 μL 20µL
16873465 150 μL 150µL
2 options
Product Code. 16883465
Brand: Proteintech 249441AP20UL
Rabbit Polyclonal Antibody
ME2 encodes a mitochondrial NAD-dependent malic enzyme, a homotetrameric protein, that catalyzes the oxidative decarboxylation of malate to pyruvate. It had previously been weakly linked to a syndrome known as Friedreich ataxia that has since been shown to be the result of mutation in a completely different gene. Certain single-nucleotide polymorphism haplotypes of this gene have been shown to increase the risk for idiopathic generalized epilepsy. Alternatively spliced transcript variants encoding different isoforms found for this gene.
Specifications
Antigen ME2
Applications Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ME2
Gene Accession No. P23368
Gene Alias Malic enzyme 2, ME2, NAD ME
Gene Symbols ME2
Host Species Rabbit
Immunogen ME2 Fusion Protein Ag20741
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4200
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG