missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ME2 Polyclonal antibody specifically detects ME2 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Dilution | Western Blot |
| Gene Alias | EC 1.1.1, EC 1.1.1.38, malate dehydrogenase, Malic enzyme 2, malic enzyme 2, NAD(+)-dependent, mitochondrial, NAD-dependent malic enzyme, mitochondrial, NAD-ME, ODS1, pyruvic-malic carboxylase |
| Research Discipline | Biologically Active Proteins |
| Primary or Secondary | Primary |
| Target Species | Human |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?