Learn More
Karl Hecht Polypropylene measuring jug with 100ml scale
Brand:  Karl Hecht 42283030
Polypropylene measuring jug with 100ml scale
Product Code. 10013931
  • ONLINE EXCLUSIVE PRICE
    48.30 NOK
    33.95 NOK / Pack of 1
    Save 14.35 NOK
    29% Off
 Restricted Item
Estimated Shipment: 21-10-2026