Learn More
MDM2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Product Code. 16875885 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16875885 150 μL 150µL
16885885 20 μL 20µL
2 options
Product Code. 16875885
Brand: Proteintech 278831AP150UL
Rabbit Polyclonal Antibody
The MDM2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MDM2. This antibody recognizes human antigen. The MDM2 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
Antigen MDM2
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Concentration 367 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 4193
Target Species Human
Isotype IgG