Learn More
VACUUBRAND™ MD 12C NT +EK Chemistry Vacuum System
Product Code. 12623315
Change view
Click to view available options
Flow Rate:
7.1/7.8 CFM
Unit Size:
Each
Product Code. Flow Rate unitSize
12623315 7.1/7.8 CFM Each
1 options
Product Code. 12623315
Brand: VACUUBRAND™ 20743500
This chemistry vacuum system has a wide range of applications like evacuation, evaporation and pumping of gases and vapors.
  • Provides a particularly high pumping speed and are ideal for higher vacuum requirements, e.g., with high boiling solvents
  • Obtains very good ultimate vacuum even with gas ballast and features high performance in a compact design
  • The consistent design for highest chemical resistance enables almost universal usage in chemistry and pharmaceutics
  • The waste vapor condenser at the outlet (EK) is highly efficient and compact, enabling a near-100% solvent recovery, efficient recycling and active protection of the environment
  • An optional separator at the inlet (AK) made of glass with protective coating retains particles and liquid droplets and can be mounted later to this system
  • Whisper-quiet and very low vibration
Order Info
Includes
TRUSTED_SUSTAINABILITY
Specifications
Max. Vacuum 1.5 torr (without Gas Ballast), 3 torr (with Gas Ballast)
Horsepower 0.59 hp
Includes Manual
Motor Speed 150 to 1800 rpm
Certifications/Compliance ATEX Conformity II 3G IIC T3 X, CE, DIN EN 1012-2:2011, DIN EN 12100:2001, DIN EN 50581:2013, DIN EN 61010-1:2011, DIN EN 61326-1:2013, IEC 61010-1:2010
Electrical Requirements 230 V 50/60 Hz
Dimensions (L x W x H) 528 x 387 x 395 mm
Model MD 12C NT +EK
Noise Level 50 dB(A)
Temperature (Metric) Storage -10°C to +60°C
Temperature (English) Max. 104°F
Temperature (Metric) Max 40°C
Temperature (Metric) Min. 10°C
Flow Rate 7.1/7.8 CFM
Frequency 50/60 Hz
Overall Height (Metric) 396.24 mm
Overall Length (Metric) 528.32 mm
Overall Width (Metric) 386.08 mm
Pump Type Diaphragm Pump
Enclosure Rating IP40
No. of Pump Heads 8
Resistance Properties Chemical-Resistant
Temperature (English) Min 50°F
Oil Type Oil-Free
No. of Stages 3
Maximum Pressure 16 psi
Voltage 230 V