Learn More
Lovibond™ MD 100 Chlorine Tablet
Product Code. 12641705
Change view
Click to view available options
Detection Range:
0 to 6 mg/L, 0 to 10 mg/L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detection Range Unit Size
12641705 0 to 6 mg/L, 0 to 10 mg/L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12641705 Supplier Lovibond™ Supplier No. 276000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Lovibond MD 100 Chlorine Tablet, Applications: Testing in the field, laboratory or plant room, Detection Range: 0-6 - 0-10mg/L, Display Type: LCD, Filling Solution Type: Reagent
Specifications
Type MD 100 Chlorine Tablet
Detection Range 0 to 6 mg/L, 0 to 10 mg/L
Display Type LCD
Filling Solution Type Reagent
Temperature (Metric) Operating 5°C to 40°C
For Use With (Application) Testing in the field, laboratory or plant room
Includes 4 Micro batteries (AAA), 3 round vials (Glass) with lids, tablet reagents and/or liquid reagents, guarantee sheet, certificate (COC), instruction manual
Warranty 5 or 10 Year
Length (Metric) 155 mm
Width (Metric) 75 mm