missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM7 Mouse, Clone: MCM7/1469, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 4176-MSM4-P0
Additional Details : Weight : 0.01000kg
Specifications
MCM7 | |
Nuclear | |
MCM7/1469 | |
Unconjugated | |
NP_005907 | |
Mouse | |
88kDa | |
Protein A/G Purified | |
RUO | |
4176 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
MCM7 | |
Recombinant human MCM7 protein fragment (aa195-319) | |
Raji or Ramos cells. Tonsil | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |