Learn More
Abnova™ MCM7 (Human) Recombinant Protein
Human MCM7 full-length ORF ( AAH09398, 1 a.a. - 389 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004176-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MALKDYALEKEKVKKFLQEFYQDDELGKKQFKYGNQLVRLAHREQVALYVDLDDVAEDDPELVDSICENARRYAKLFADAVQELLPQYKEREVVNKDVLDVYIEHRLMMEQRSRDPGMVRSPQNQYPAELMRRFELYFQGPSSSKPRVIREVRADSVGKLVTVRGIVTRVSEVKPKMVVATYTCDQCGAETYQPIQSPTFMPLIMCPSQECQTNRSGGRLYLQTRGSRFIKFQEMKMQEHSDQVPVGNIPRSITVLVEGENTRIAQPGDHVSVTGIFLPILRTGFRQVVQGLLSETYLEAHRIVKMNKSEDDESGAGELTREELRQIADVIFATVRELVSGGRSVRFSEAEQRCVSRGFTPAQFQAALDEYEELNVWQVNASRTRITFV
Specifications
AAH09398 | |
minichromosome maintenance complex component 7 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MCM7 | |
GST |
4176 | |
Wheat germ expression system | |
25 μg | |
CDABP0042, CDC47, MCM2, P1.1-MCM3, P1CDC47, P85MCM, PNAS-146 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |