Learn More
Abnova™ MCM7 (Human) Recombinant Protein
Product Code. 16127671
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16127671
Brand: Abnova™ H00004176P01.10ug
Human MCM7 full-length ORF ( AAH09398, 1 a.a. - 389 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: MALKDYALEKEKVKKFLQEFYQDDELGKKQFKYGNQLVRLAHREQVALYVDLDDVAEDDPELVDSICENARRYAKLFADAVQELLPQYKEREVVNKDVLDVYIEHRLMMEQRSRDPGMVRSPQNQYPAELMRRFELYFQGPSSSKPRVIREVRADSVGKLVTVRGIVTRVSEVKPKMVVATYTCDQCGAETYQPIQSPTFMPLIMCPSQECQTNRSGGRLYLQTRGSRFIKFQEMKMQEHSDQVPVGNIPRSITVLVEGENTRIAQPGDHVSVTGIFLPILRTGFRQVVQGLLSETYLEAHRIVKMNKSEDDESGAGELTREELRQIADVIFATVRELVSGGRSVRFSEAEQRCVSRGFTPAQFQAALDEYEELNVWQVNASRTRITFV
Specifications
Accession Number AAH09398
Gene ID (Entrez) 4176
Name minichromosome maintenance complex component 7
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias CDABP0042, CDC47, MCM2, P1.1-MCM3, P1CDC47, P85MCM, PNAS-146
Gene Symbol MCM7
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer