missing translation for 'onlineSavingsMsg'
Learn More

MCM2 Mouse anti-Human, Clone: 1F4D10, Proteintech

Product Code. 16837865 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16837865 150 μL 150µL
16847865 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16837865

Brand: Proteintech 662041IG150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MCM2 (Mini chromosome maintece protein 2) is involved in regulating DNA replication. MCM2 (also called CDCL1, mitotin and BM28), is a human nuclear protein that is crucial in the cell cycle, being involved in the onset of DNA replication and cell division. It is similar to members of the family of early S-phase proteins. MCM2 has been mapped to 3q21. From its localization, CDCL1 became a candidate for an oncogene affected by chromosomal breaks in acute myeloid leukemia (AML).

Specifications

Antigen MCM2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1F4D10
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MCM2
Gene Accession No. P49736
Gene Alias BM28, CCNL1, cdc19, CDCL1, D3S3194, KIAA0030, MCM2, MITOTIN, Nuclear protein BM28
Gene Symbols MCM2
Host Species Mouse
Immunogen MCM2 Fusion Protein Ag0798
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4171
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.