Learn More
Lovibond™ MC500 Colorimeter Replacement Reagents: Chlorine
For use with MC500 multi-parameter colorimeter
Brand: Lovibond™ 517711BT
Additional Details : Weight : 0.30000kg
Beschreibung
For use with MC500 multi-parameter colorimeter
- Parameter: Chlorine
- Method: DPD
Spezifikation
Water quality testing | |
DPD No. 1 tablets (100 pack), DPD No. 3 tablets (100 pack) | |
0.01 to 6mg/L | |
Chlorine Reagent Set |
MC500 multi-parameter colorimeter | |
100 | |
DPD |