Learn More
Bibby Scientific™ MC227 1-Way Heating Controller
Product Code. 11502724
Change view
Click to view available options
Diameter (Metric):
12.5mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11502724 12.5mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11502724 Supplier Bibby Scientific™ Supplier No. 3622300
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
In Stock
Specifications
Diameter (Metric) 12.5mm
Depth (Metric) 115 mm
Dimensions (L x W x H) 115 x 120 x 80mm
Height (Metric) 80 mm
Voltage 230 V
Electrical Requirements 230V, 50/ 60 Hz, 2300W
Model MC227
Power Supply Details 1w
Wattage 2300w
Weight (Metric) 0.82Kg
Width (Metric) 120mm
Product Type 1-way Heating Controller
Frequency 50/60 Hz