Learn More
MC1568, MedChemExpress
Product Code. 16487186
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16487186
Brand: Medchem Express HY16914
MedChemExpress MC1568 is a selective class II (IIa) histone deacetylas (HDAC II) inhibitor, used for cancer research.
MC1568 is a selective class II (IIa) histone deacetylas (HDAC II) inhibitor, used for cancer research.
Specifications
Chemical Name or Material MC1568
CAS 852475-26-4
Color Red
Purity Grade Notes Research
Molecular Formula C17H15FN2O3
Quantity 100 mg
Solubility Information DMSO : 18.5 mg/mL (58.86 mM; Need ultrasonic and warming)
SMILES O=C(NO)/C=C/C1=CC=C(/C=C/C(C2=CC=CC(F)=C2)=O)N1C
Molecular Weight (g/mol) 314.31
Formula Weight 314.31
Percent Purity 96.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.