Learn More
MBP146-78, MedChemExpress
Product Code. 16499926
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
Product Code. 16499926
Brand: Medchem Express HY101525
MedChemExpress MBP146-78 is a potent and selective inhibitor of cGMP dependent protein kinases.
MBP146-78 is a potent and selective inhibitor of cGMP dependent protein kinases.
Specifications
Chemical Name or Material MBP146-78
CAS 188343-77-3
Color White
Purity Grade Notes Research
Molecular Formula C21H22FN3
Quantity 50 mg
Solubility Information DMSO : 7.69 mg/mL (22.93 mM; Need ultrasonic)
SMILES CN1CCC(C2=CC(C3=CC=NC=C3)=C(C4=CC=C(F)C=C4)N2)CC1
Molecular Weight (g/mol) 335.42
Formula Weight 335.42
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.