Learn More
MBL™ Type 4 Class B Serological Graduated Pipettes
Manufactured from soda-lime glass with robust tooled jets
Brand: MBL PRA842S
233.70 NOK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.19000kg
Description
- Calibrated to deliver from any graduation line down to zero at the jet with the last drop expelled by blowing
- Tooled suction ends accept cotton-wool plug
- Graduations in permanent amber stain
Specifications
ISO 835 | |
Yes | |
5/Pk. | |
2 mL |
Soda Lime Glass | |
0.02 mL | |
±0.02 mL | |
Serological Pipette |