Learn More
MBL™ Dropping Pipettes for Soda-Lime Reagent Bottles
Click to view available options
Description:
14/15 Joint
14/23 Joint
19/26 Joint
For Use With (Equipment):
100 mL Bottle
250 mL Bottle
50 mL Bottle
Unit Size:
Pack of 5
Description
Manufactured from clear soda-lime glass
- With integral ground glass cone on pipette
- Supplied with rubber teats
- Bottles available separately
Specifications
Specifications
| Description | 14/15 Joint |
| Material | Soda Lime Glass |
| Includes | Dropping pipettes with silicone teat |
| Type | Pasteur Pipet |
| For Use With (Equipment) | 50 mL Bottle |