Learn More
MBL™ Borosilicate Glass Class A Volumetric Flask with Stopper
Product Code. 11381915
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
5 L
Height (Metric):
70 mm
90 mm
110 mm
140 mm
170 mm
210 mm
220 mm
260 mm
300 mm
370 mm
475 mm
Unit Size:
Each
Pack of 2
Pack of 5
Closure Size:
10/19
12/21
14/23
19/26
24/29
29/32
34/35
Product Code. 11381915
Brand: MBL FRD166
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Complies with ISO 1042 and DIN 12664. MBL™ Borosilicate Glass Class A Volumetric Flask with Stopper are manufactured from chemically resistant borosilicate glass.
  • Finely calibrated to Class A tolerances
  • Each flask is laser etched with an individual serial number, batch code and date of manufacture
  • Conformity 'H' certified
  • Inscriptions in blue enamel
  • Supplied with polyethylene stopper
  • Batch certificate available to download online
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Class A, ISO 1042, DIN 12664, Conformity 'H' certified
Closure Material Polyethylene
Description Blue enamel inscriptions
Capacity (Metric) 10 mL
Material Borosilicate Glass
Height (Metric) 90 mm
Tolerance ±0.025 mL
Graduated Yes
Class A