Learn More
Axygen™ Maxymum Recovery™ 200 μL Universal Pipetter Tips: 200 μL, Beveled
Product Code. 12655445 Shop All Axygen Scientific Products
Change view
Click to view available options
Format:
Bulk
Hinged Rack
Stacked Rack
Unit Size:
Pack of 1000
Pack of 20000
Pack of 4800
Pack of 960
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format unitSize
12655445 Bulk Pack of 1000
12561358 Bulk Pack of 20000
12639535 Hinged Rack Pack of 960
12571288 Stacked Rack Pack of 4800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12655445 Supplier Axygen™ Supplier No. T200CL
Only null left
Add to basket
Add to basket
Designed to fit a wide variety of single and multi-channel pipettes
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Maxymum Recovery technology for ultra-low retention
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 1 to 200 μL
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, DNA, and PCR inhibitor Free
Format Bulk
Tip Style Beveled, Low Retention
Packaging Bulk; 1,000 Tips/Pk, 20 Packs/Cs
For Use With (Equipment) Universal Fit
Product Line Maxymum Recovery