Learn More
Thermo Scientific™ MaxQ™ Shaker Universal Platforms
Product Code. 15599843 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Dual Stacked Universal Platform w/o Clamps
Platform w/o Clamps
Stacking Adapter
Universal Platform w/o Clamps
For Use With (Equipment):
MaxQ 2000
MaxQ 2000, 2506, 2508, 4000, 6000
MaxQ 2000, 2506, 2508, 4450
MaxQ 2000, 6000
MaxQ 3000
MaxQ 3000, 5000
MaxQ 30100, MaxQ 30110 Platform
MaxQ 7000
Dimensions (L x W):
11 x 13 in.
13 x 11 in.
18 x 18 in.
18 x 30 in.
24 x 36 in.
Unit Size:
Each
Product Code. Product Type For Use With (Equipment) Dimensions (L x W) unitSize
15599843 Dual Stacked Universal Platform w/o Clamps MaxQ 2000 11 x 13 in. Each
10369032 Platform w/o Clamps MaxQ 2000, 2506, 2508, 4450 11 x 13 in. Each
10275852 Platform w/o Clamps MaxQ 7000 11 x 13 in. Each
10798914 Universal Platform w/o Clamps MaxQ 2000, 2506, 2508, 4000, 6000 18 x 18 in. Each
10061552 Universal Platform w/o Clamps MaxQ 3000, 5000 18 x 30 in. Each
10071552 Universal Platform w/o Clamps MaxQ 3000 24 x 36 in. Each
12546299 Dual Stacked Universal Platform w/o Clamps MaxQ 2000, 6000 18 x 18 in. Each
10689623 Stacking Adapter MaxQ 30100, MaxQ 30110 Platform 13 x 11 in. Each
8 options
Product Code. 15599843
Brand: Thermo Scientific™ 30125
Only null left
Add to basket
Add to basket
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms.
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms. With exceptional innovation, reliability and accuracy, these platforms are the optimal choice for a busy lab.
For use with MaxQ Shakers as indicated. Choose the platform that best suits your needs.
Ordering Information:
Purchase clamps separately.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Dual Stacked Universal Platform w/o Clamps
Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC)
Dimensions (L x W) 11 x 13 in.
Length (English) Platform 11 in.
Length (Metric) Platform 28 cm
Width (English) Platform 13 in.
Width (Metric) Platform 33 cm
For Use With (Equipment) MaxQ 2000