Learn More
Thermo Scientific™ Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Product Code. 10041592 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Adhesive Mat applicator
Dual Height Adjustable Clamp
Flask Clamp
Flask Clamp Kit
Media Bottle Clamp
Microplate/Deep-Well Plate Clamp
Separatory Funnel Clamp
Single Height Adjustable Clamp
Vessel Clamp
Holds:
1 L Erlenmeyer Flask
10 mL Erlenmeyer Flask
1000 mL Square Media Bottle
125 mL Erlenmeyer Flask
125 mL Square Media Bottle
2 L Erlenmeyer Flask
25 mL Erlenmeyer Flask
250 mL Erlenmeyer Flask
250 mL Square Media Bottle
2500 mL Low-form Culture Flask
2800 mL Fernbach Flask
300 mL Erlenmeyer Flask
4 L Erlenmeyer Flask
5 L Erlenmeyer Flask
50 mL Erlenmeyer Flask
500 mL Erlenmeyer Flask
500 mL Square Media Bottle
Flasks
Microplate/Deep-Well Plate
Separatory Funnel
Single Vessel
Vessel
Vessels
For Use With (Equipment):
Solaris Open Air and Temperature Controlled Shakers, MaxQ 6000 Shakers, MaxQ 416 HP Shakers
Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
Unit Size:
Each
Product Code. Product Type Holds For Use With (Equipment) unitSize
10041592 Flask Clamp 2 L Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
12354263 Flask Clamp Kit Flasks Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
10484302 Flask Clamp 10 mL Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
10153022 Flask Clamp 25 mL Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
10549912 Flask Clamp 50 mL Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
10001572 Flask Clamp 125 mL Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
10326632 Flask Clamp 250 mL Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
10685663 Flask Clamp 300 mL Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
10494492 Flask Clamp 500 mL Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
10404502 Flask Clamp 1 L Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
10414692 Flask Clamp 4 L Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
10336632 Flask Clamp 5 L Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
10579532 Flask Clamp 2500 mL Low-form Culture Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
10579722 Flask Clamp 2800 mL Fernbach Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
10396722 Microplate/Deep-Well Plate Clamp Microplate/Deep-Well Plate Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
17831895 Vessel Clamp Single Vessel Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
17881875 Dual Height Adjustable Clamp Vessel Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
17821895 Dual Height Adjustable Clamp Vessel Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
17871875 Single Height Adjustable Clamp Vessel Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
17841895 Media Bottle Clamp 125 mL Square Media Bottle Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
17891875 Media Bottle Clamp 250 mL Square Media Bottle Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
17861895 Media Bottle Clamp 500 mL Square Media Bottle Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
17811885 Media Bottle Clamp 1000 mL Square Media Bottle Solaris Open Air and Temperature Controlled Shakers, MaxQ 6000 Shakers, MaxQ 416 HP Shakers Each
17821905 Separatory Funnel Clamp Separatory Funnel Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
17841885 Adhesive Mat applicator Vessels Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers Each
25 options
Product Code. 10041592
Brand: Thermo Scientific™ 30158
Only null left
Add to basket
Add to basket
Mix and match these shaker platform clamps, mats and racks, available in a variety of sizes, to suit individual needs in the laboratory.
Mix and match Thermo Scientific™ MaxQ™ Shaker Universal Clamps, available in a variety of sizes, to suit individual needs in the laboratory. Compatible with MaxQ shaker platforms.
Clamps are sold as individual units.
TRUSTED_SUSTAINABILITY
Specifications
Holds 2 L Erlenmeyer Flask
Product Type Flask Clamp
For Use With (Equipment) Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers