Learn More
- Home
- Products
- Shakers
- Shaker Accessories
- Thermo Scientific™ Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Thermo Scientific™ Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Product Code. 10001572 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Adhesive Mat applicator
Dual Height Adjustable Clamp
Flask Clamp
Flask Clamp Kit
Media Bottle Clamp
Microplate/Deep-Well Plate Clamp
Separatory Funnel Clamp
Single Height Adjustable Clamp
Vessel Clamp
Holds:
1 L Erlenmeyer Flask
10 mL Erlenmeyer Flask
1000 mL Square Media Bottle
125 mL Erlenmeyer Flask
125 mL Square Media Bottle
2 L Erlenmeyer Flask
25 mL Erlenmeyer Flask
250 mL Erlenmeyer Flask
250 mL Square Media Bottle
2500 mL Low-form Culture Flask
2800 mL Fernbach Flask
300 mL Erlenmeyer Flask
4 L Erlenmeyer Flask
5 L Erlenmeyer Flask
50 mL Erlenmeyer Flask
500 mL Erlenmeyer Flask
500 mL Square Media Bottle
Flasks
Microplate/Deep-Well Plate
Separatory Funnel
Single Vessel
Vessel
Vessels
For Use With (Equipment):
Solaris Open Air and Temperature Controlled Shakers, MaxQ 6000 Shakers, MaxQ 416 HP Shakers
Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
Unit Size:
Each
25 product options available for selection
| Product Code. | Product Type | Holds | For Use With (Equipment) | Unit Size |
|---|---|---|---|---|
| 10001572 | Flask Clamp | 125 mL Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 12354263 | Flask Clamp Kit | Flasks | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 10484302 | Flask Clamp | 10 mL Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 10153022 | Flask Clamp | 25 mL Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 10549912 | Flask Clamp | 50 mL Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 10326632 | Flask Clamp | 250 mL Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 10685663 | Flask Clamp | 300 mL Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 10494492 | Flask Clamp | 500 mL Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 10404502 | Flask Clamp | 1 L Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 10041592 | Flask Clamp | 2 L Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 10414692 | Flask Clamp | 4 L Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 10336632 | Flask Clamp | 5 L Erlenmeyer Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 10579532 | Flask Clamp | 2500 mL Low-form Culture Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 10579722 | Flask Clamp | 2800 mL Fernbach Flask | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 10396722 | Microplate/Deep-Well Plate Clamp | Microplate/Deep-Well Plate | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 17831895 | Vessel Clamp | Single Vessel | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 17881875 | Dual Height Adjustable Clamp | Vessel | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 17821895 | Dual Height Adjustable Clamp | Vessel | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 17871875 | Single Height Adjustable Clamp | Vessel | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 17841895 | Media Bottle Clamp | 125 mL Square Media Bottle | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 17891875 | Media Bottle Clamp | 250 mL Square Media Bottle | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 17861895 | Media Bottle Clamp | 500 mL Square Media Bottle | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 17811885 | Media Bottle Clamp | 1000 mL Square Media Bottle | Solaris Open Air and Temperature Controlled Shakers, MaxQ 6000 Shakers, MaxQ 416 HP Shakers | Each |
| 17821905 | Separatory Funnel Clamp | Separatory Funnel | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
| 17841885 | Adhesive Mat applicator | Vessels | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
Mix and match Thermo Scientific™ MaxQ™ Shaker Universal Clamps, available in a variety of sizes, to suit individual needs in the laboratory. Compatible with MaxQ shaker platforms.
Clamps are sold as individual units.
Specifications
Specifications
| Holds | 125 mL Erlenmeyer Flask |
| Product Type | Flask Clamp |
| For Use With (Equipment) | Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers |