Learn More
Thermo Scientific™ MaxQ™ HP Orbital Shakers Universal Platforms
Product Code. 11801271 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
MaxQ 8000
MaxQ HP 416
MaxQ HP 416/420
MaxQ HP 430
MaxQ HP 430/435/436/480/481
Dimensions (L x W):
18 x 18 in.
18 x 24 in.
18 x 29.5 in.
24 x 36 in.
Unit Size:
Each
Product Code. For Use With (Equipment) Dimensions (L x W) unitSize
11801271 MaxQ HP 430/435/436/480/481 18 x 29.5 in. Each
11836763 MaxQ HP 416/420 18 x 18 in. Each
10115432 MaxQ HP 430 24 x 36 in. Each
10024102 MaxQ HP 416 18 x 24 in. Each
10594093 MaxQ 8000 18 x 29.5 in. Each
5 options
Product Code. 11801271
Brand: Thermo Scientific™ 238001
Only null left
Add to basket
Add to basket
Mix and match different size labware on a single platform with these orbital shakers universal platforms.
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ HP Orbital Shakers Universal Platforms.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Universal Platform
Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC)
Dimensions (L x W) 18 x 29.5 in.
Length (English) Platform 29.5 in.
Length (Metric) Platform 74.9 cm
Width (English) Platform 18 in.
Width (Metric) Platform 45.7 cm
Description Universal Platform without Clamps
For Use With (Equipment) MaxQ HP 430/435/436/480/481