Learn More
Thermo Scientific™ Oxoid™ Maximum Recovery Diluent
Description
Achieve maximum recovery of microorganisms during dilution with this protective, isotonic Thermo Scientific™ Oxoid™ Maximum Recovery Diluent.
The presence of peptone in the diluent provides viable counts to ensure accurate quantitative procedures are performed. The isotonic strength of the diluent ensures recovery of organisms from various sources which may be vulnerable in distilled water or aqueous suspensions. The low peptone concentration in the diluent reduces organism multiplication in the sample for at least 45 minutes during the dilution stage1,2
Maintain the viability of organisms during dilution process with Maximum Recovery Diluent with low levels of peptone.
- Allows accurate quantitative procedures to be performed with minimal reduction in viable count.
- Reduces organism multiplication in the sample for at least one hour during the dilution stage.
- Ready-to-use.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Color | Colorless, clear |
| Content And Storage | 2 to 25°C |
| Description | Maximum Recovery Diluent |
| Format | Universal bottles |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar |
| Fill Weight | 9.0g + 0.3g |
| Packaging Type | Twenty four universal bottles with screw cap closures in a box |
| Quantity | 24/Pk. |