Learn More
Thermo Scientific™ Maxima H Minus Reverse Transcriptase Promotion
Product Code. 15777496 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10,000 U
2000 U
Unit Size:
10000 units
2000 units
Product Code. Quantity unitSize
15777496 10,000 U 10000 units
15767496 2000 U 2000 units
2 options
Product Code. 15777496
Brand: Thermo Scientific™ EP0752
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A highly thermostable, high processivity reverse transcriptase lacking RNase H activity
  • Thermostable - 90% active after incubation at 50°C for 60 minutes in a reaction mixture.
  • Active up to 65°C
  • High yields of full-length cDNA up to 20 kb
  • High sensitivity - reproducible cDNA synthesis from a wide range of starting total RNA amounts (1 pg to 5 μg)
  • Efficient - complete cDNA synthesis in 15 to 30 minutes
  • Increased resistance to common reaction inhibitors
  • Incorporates modified nucleotides
TRUSTED_SUSTAINABILITY
Specifications
Concentration 5X
Composition 250 mM Tris-HCl buffer (pH 8.3 at 25°C), 37 5mM KCl, 15 mM MgCl2, 50 mM DTT
Content And Storage -20°C
Quantity 10,000 U
For Use With (Application) cDNA Synthesis, RT-PCR, Gene Expression