Learn More
3M™ Maxim™ RAS Ballistic Safety Glasses
Click to view available options
:
Lens Tint: Clear
Description
- Safety spectacles that exceed the Military VO test regimen for high velocity impact
- RAS coating 5 times more scratch resistance than generic hard coats
- RAS coating fulfils the EN 166 requirements for K marking
- Spectacles with ventilation channel across the lens to minimize misting
- Soft, adjustable nose bridge and a soft-touch brow guard
- Adjustable temple lengths and pantoscopic (angle) adjustments
- Design provides excellent coverage and good field of vision
- Low-profile design increases compatibility with helmets and hearing protection
Specifications
Specifications
| Certifications/Compliance | EN 166: 2001 |
| Lens Coating | Scratch-resistant |
| Lens Tint | Clear |
| Product Type | Safety glasses |
| For Use With (Application) | Construction, metallurgy |
| Frame Color | Black |
| Lens Material | Polycarbonate |