Learn More
Honeywell™ Maxi Welder Cut Gloves
Product Code. 12667513
Change view
Click to view available options
Size: 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12667513 Size: 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 12667513 Supplier Honeywell™ Supplier No. 205859010
Quantity Price
1 533.32 NOK / Pack of 1
10 506.65 NOK / Pack of 1
Estimated Shipment: 14-12-2026
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Honeywell™ Maxi Welder Cut Gloves for MIG and arc welding when handling dry, greasy, or sharp parts.
  • Cowhide leather/Kevlar™ glove
  • Gunn cut, winged thumb with yoke, seamless forefinger, gauntlet type
  • Silicon coated Kevlar™ palm, lined with cotton
  • Water-repellent cowhide grain leather back
  • Elastic on the back
  • 150 mm split leather cuff
  • Stitch with Kevlar™ thread
  • Thickness leather only 0.9/1.1 mm
  • The hide leather is ideal for welders' gloves
  • The US assembly, winged thumb and flexible materials used make this glove anatomical, thus providing excellent comfort for use
  • The Kevlar™ fleece lining with its cotton lining increases the glove's comfort and heat insulation
  • Split leather treated against the heat for excellent resistance to contact and radiant heat
  • The water repellent grain leather back is ideal for work in a damp atmosphere
  • The black Kevlar™ silicone palm protects the glove against cuts and thermal hazards
  • The split leather cuff protects a large part of the forearm against mechanical hazards
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE CATEGORY : II Intermediary risksIn accordance with STANDARDS :EN 420-03 EEC Basic standardsEN 388-03 Mechanical hazardsEN 407-04 Heat hazardsEN 12477-2000 Welding hazards
Description Silicon coated Dupont™ Kevlar™ aramid fiber palm
Color Black, Yellow, White
Size 10
Thickness (Metric) 0.9 to 1.1 mm
Length (Metric) 38.5 cm