Learn More
SICCO Maxi 2-Desiccator Antistatic
Click to view available options
Volume (Metric):
156 L
Unit Size:
Each
Description
- Aluminium frame with static dissipative panels made of polycarbonate, two compartments and two doors, including four shelves made of stainless steel, two trays and two hygrometers, four casters with static dissipative tread, usable with up to 17 shelves per compartment.
- All materials used can discharge electrostatic charging by means of a grounding cable which can be connected on the back side (connecting thread M6).
- Reinforced aluminium frame with panels made of antistatic polycarbonate
- Door with magnetic catch and circular rubber seal
- Four casters (two of the casters with brakes)
- Easy to read electronic hygrometer
- Variable height shelves made of stainless steel
- Telescopic shelves
- Connection for grounding cable
- Tray for desiccant
Specifications
Specifications
| Autoclavable | Autoclavable |
| Cabinet Style | Front-loading |
| Volume (Metric) | 156 L |
| Dimensions (D x W x H) Interior | 560 x 1150 x 580 mm |
| Weight (Metric) | 34 kg |