Learn More
Fisherbrand™ Max Mono II Monocular Biological Microscope
Click to view available options
Eyepiece:
10X/18 mm
Unit Size:
Each
Description
- Reversed 4-position turret for easy access to stage
- With carrying handles
- Full mechanical stage
- Pre-centered Abbe condenser
- With three achromatic objectives – 4x, 10x, 60x
- 20W illumination and comes with a blue filter and dust cover
Specifications
Specifications
| Condenser Type | Abbe |
| Eyepiece | 10X/18 mm |
| Head Style | Rotating |
| Lighting Type | Halogen |
| Model | Max Mono II |
| Objective Types | Achromatic |
| Electrical Requirements | 220/240 V 50/60 Hz |
| Voltage | 220/240 V |
| Frequency | 50/60 Hz |