Learn More
IKA™ MATRIX ThermoCover
Product Code. 16238701 Shop All IKA Products
Click to view available options
For Use With (Equipment):
MATRIX Orbital Delta F1.5, MATRIX Orbital Delta FP, MATRIX Orbital Delta F0.5, MATRIX Orbital Delta F2.0, MATRIX Orbital Delta Plus, MATRIX Delta Plus
Unit Size:
Each
Product Code. 16238701
Brand: IKA™ 0030001244
Only null left
Add to basket
Add to basket
The heatable ThermoCover prevents condensation on the vial lid and wall. At the same time it enables constant reaction conditions.
Suitable for: MATRIX Orbital Delta F1.5 MATRIX Orbital Delta FP MATRIX Orbital Delta F0.5 MATRIX Orbital Delta F2.0 MATRIX Orbital Delta plus MATRIX Delta plus
TRUSTED_SUSTAINABILITY
Specifications
Product Type Heatable ThermoCover
Dimensions (L x W x H) 190 x 180 x 120 mm
For Use With (Application) Preventing the formation of condensate on vessel walls/vessel lids
For Use With (Equipment) MATRIX Orbital Delta F1.5, MATRIX Orbital Delta FP, MATRIX Orbital Delta F0.5, MATRIX Orbital Delta F2.0, MATRIX Orbital Delta Plus, MATRIX Delta Plus