Learn More
Thermo Scientific™ Matrix™ Pipette Tips In Ecotips Refill Packaging System
Product Code. 10314833
Change view
Click to view available options
Description:
ECOTIPS
TallTip ECOTIPS
Volume (Metric):
250 μL
1250 μL
Unit Size:
Pack of 960
Product Code. Description Volume (Metric) unitSize
10314833 ECOTIPS 250 μL Pack of 960
10591135 ECOTIPS 1250 μL Pack of 960
10735014 TallTip ECOTIPS 1250 μL Pack of 960
3 options
Product Code. 10314833
Brand: Thermo Scientific™ 7156
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Process samples quickly with fast rack reloading in an environmentally friendly, space-saving design with this unique tip refill system.
Process samples quickly with fast rack reloading in an environmentally friendly, space-saving design with the Thermo Scientific™ Matrix™ Pipette Tips In Ecotips Refill Packaging System, which reduces plastic waste by eliminating the need for a new tip box for each rack of tips. This system is 100% recyclable and each component is marked with appropriate recycling symbols. Trays of tips are nested in easily accessible packaging that takes up very little space and eliminates the need to dispose of a tip box each time a rack is emptied.
  • Compact alternative to standard racked tips
  • Quick, efficient refill; reloading empty tip boxes is quick and easy
  • Made from 100% virgin polypropylene; insert trays and boxes come from our own recycled materials
  • Saves bench, shelf and closet space; refills fit easily in a drawer, on a shelf or on the bench.
  • Allows for quick and efficient transfer of tips from packaging to empty tip box
  • Finger tabs on tip trays for easy removal of empty inserts from each tip box
  • Eliminates the need for mechanical towers, clumsy stacks of tips or other transfer mechanisms
TRUSTED_SUSTAINABILITY
Specifications
Description ECOTIPS
Volume (Metric) 250 μL
Filtered Non-filtered
Sterility Non-sterile
Format Refill
For Use With (Equipment) Thermo Scientific Matrix
Product Line Ecotips, Matrix