Learn More
Matrix oil, 20 centistoke hydrocarbon oil, Thermo Scientific Chemicals
Product Code. 15451947
Change view
Click to view available options
Quantity:
470 mL
3.8 L
Unit Size:
3.8 L
470mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15451947 470 mL 470mL
15461947 3.8 L 3.8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15451947 Supplier Thermo Scientific Alfa Aesar Supplier No. 044012.#2
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 64742-65-0
MDL Number MFCD00147539
Specifications
Odor Petroleum-like
Physical Form Liquid
Quantity 470 mL
Chemical Name or Material Matrix oil, 20 centistoke hydrocarbon oil