missing translation for 'onlineSavingsMsg'
Learn More

IKA™ MATRIX Cryo attachment

Product Code. 16238691 Shop All IKA Products
Click to view available options
For Use With (Equipment):
MATRIX Orbital Delta Plus, MATRIX Delta Plus
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16238691

Brand: IKA™ 0030000848

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The MATRIX Cryo attachment is suitable for 24 x Cryo vials (1.5 - 2.0 ml, outer diameter max. 12.5 mm). The ideal attachment design ensures an optimal temperature transfer and thus guarantees valid results.

The thermoshakers MATRIX Orbital Delta plus and MATRIX Delta plus automatically recognize, which attachment is connected to the device via an interface in the locking mechanism. The attachment can be changed quickly and easily with an ergonomic lever. As part of IKA's Integrated Safety Concept, the insulating plastic coating of the attachment ensures user safety, as high temperatures are not transferred to the frame. Speed max.: 2000 rpm Temperature max.: 110 °C Suitable for: MATRIX Orbital Delta plus MATRIX Delta plus
TRUSTED_SUSTAINABILITY

Specifications

Holds Cryo Reaction Vessels
Tube Capacity 1.5/2 mL
Product Type Tube Matrix Attachment
Dimensions (L x W x H) 150 x 141 x 50 mm
For Use With (Application) Samples with even the smallest volume, such as blood samples, pharmaceutical agents/DNA/RNA samples
For Use With (Equipment) MATRIX Orbital Delta Plus, MATRIX Delta Plus
No. of Positions 24
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.