Learn More
Corning™ Matrigel™ Matrix for Organoid Culture
BIOPHARMA
Product Code. 16491434
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491434 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16491434 Supplier Corning™ Supplier No. 356255
 Restricted Item
Estimated Shipment: 17-11-2026
Corning™ Matrigel™ Matrix for Organoid Culture is a solubilized basement membrane preparation that has been optimized for organoid culture.
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
TRUSTED_SUSTAINABILITY
Specifications
Concentration Lot dependent
Product Type Stem Cell Media
For Use With (Application) Organoid Culture and workflow
Quantity 10 mL
Source Engelbreth-Holm-Swarm (EHS) mouse tumor
Content And Storage -20°C