Learn More
Corning™ Matrigel™ Matrix-3D Plate, 384-well
Description
- A high-throughput pre-coated format that contains Matrigel™ and can be used for drug discovery applications with 3D cell cultures such as organoids, spheroids or tumoroids
- Extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors
- Each lot is verified for intraplate consistency
- Tested and found negative for the presence of bacteria and fungi
- Easy and convenient as Matrigel™ is pre-dispensed into each well
- Formats provide intra-plate consistency
- Allows for sandwich/overlay and embedded 3D application workflows
Specifications
Specifications
| Barcode | No Barcode |
| Breakapart | No |
| Lid | With Lid |
| Content And Storage | Store at -20°C |
| Autoclavable | Not Autoclavable |
| Color | Black |
| Disposable | Single-use |
| Material | Polystyrene |
| No. of Wells | 384 |
| Surface Treatment | Matrigel Matrix |
| Show More |