missing translation for 'onlineSavingsMsg'
Learn More

MATK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10082-2-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16846233

  • 3828.53 NOK / 150µL
Estimated Shipment: 17-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

All members of the Src gene family of tyrosine kinases are characterized by a carboxy terminal domain tyrosine, Y527 in the case of Src p60, which is highly phosphorylated in the inactive form of the enzyme, while phosphorylated to a much lesser extent when the enzyme is active. For instance, a mutant of c-Src, in which Y527 is replaced by phenylalanine, is transforming and displays 5 to 10-fold elevated kinase activity compared to its normal counterpart. Csk has been identified as a Src related tyrosine kinase having both SH2 and SH3 domains and a catalytic domain but lacking sequences amino terminal to the SH3 domain as well as the carboxy terminal regulatory sequences. Csk phosphorylates Src on Y527 and also down regulates Fyn, Fyn and Lck by tyrosine phosphorylation of carboxy terminal regulatory sites. A Csk-like protein-tyrosine kinase of mouse cell origin (Ctk), also designated Ntk, and its human homolog, Lsk, have also been described.
Specifications

Specifications

MATK
Polyclonal
Unconjugated
MATK
CHK, CTK, HHYLTK, HYL, HYLTK, Lsk, MATK, Protein kinase HYL, Tyrosine protein kinase CTK
Rabbit
Antigen Affinity Chromatography
RUO
17179, 4145, 60450
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P41242, P41243, P42679
Matk
MATK Fusion Protein Ag0128
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers