missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MATK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10082-2-AP-20UL
This item is not returnable.
View return policy
Description
All members of the Src gene family of tyrosine kinases are characterized by a carboxy terminal domain tyrosine, Y527 in the case of Src p60, which is highly phosphorylated in the inactive form of the enzyme, while phosphorylated to a much lesser extent when the enzyme is active. For instance, a mutant of c-Src, in which Y527 is replaced by phenylalanine, is transforming and displays 5 to 10-fold elevated kinase activity compared to its normal counterpart. Csk has been identified as a Src related tyrosine kinase having both SH2 and SH3 domains and a catalytic domain but lacking sequences amino terminal to the SH3 domain as well as the carboxy terminal regulatory sequences. Csk phosphorylates Src on Y527 and also down regulates Fyn, Fyn and Lck by tyrosine phosphorylation of carboxy terminal regulatory sites. A Csk-like protein-tyrosine kinase of mouse cell origin (Ctk), also designated Ntk, and its human homolog, Lsk, have also been described.Specifications
| MATK | |
| Polyclonal | |
| Unconjugated | |
| MATK | |
| CHK, CTK, HHYLTK, HYL, HYLTK, Lsk, MATK, Protein kinase HYL, Tyrosine protein kinase CTK | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17179, 4145, 60450 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P41242, P41243, P42679 | |
| Matk | |
| MATK Fusion Protein Ag0128 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction