Description
- Suit the needs of frequently visited washrooms with this Advanced quality paper towel roll.
- Reduce waste and boost hygiene with one-at-a-time dispensing from a compatible Tork paper towel dispenser.
- Balance cost-effectiveness with great hand-drying performance with these advanced quality hand towels.
- Commit to better environmental practices with this EU Ecolabel certified product.
Specifications
Specifications
| Product Type | Hand Towel Roll |
| Length (English) | 164.04 yd. |
| Length (Metric) | 150 m |
| Width (English) | 8.27 in. |
| Width (Metric) | 21 cm |
| Material | Paper |
| Certifications/Compliance | EU Ecolabel, ISO 50001, FSC Chain-Of-Custody, OHSAS 18001, HACCP, EMAS, FSC Mix Virgin and Rec.fibre, ISO 9001, ISO 14001 |
| Color | Blue |
| Packaging Quantity | 6/Cs. |
| Ply | 2-ply |
| Show More |