missing translation for 'onlineSavingsMsg'
Learn More

MATE 2 Rabbit anti-Canine, Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26873-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16885205

  • 1281.92 NOK / 20µL
Estimated Shipment: 14-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

S47A2 belongs to a family of transporters involved in excretion of toxic electrolytes, both endogenous and exogenous, through urine and bile. This transporter family shares homology with the bacterial MATE (multidrug and toxin extrusion) protein family responsible for drug resistance. This gene is one of two members of the MATE transporter family located near each other on chromosome 17.
Specifications

Specifications

MATE 2
Polyclonal
Unconjugated
SLC47A2
hMATE 2, MATE 2, MATE2, MATE2 B, MATE2 K, SLC47A2
Rabbit
Antigen Affinity Chromatography
RUO
146802, 489527
-20°C
Liquid
Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q86VL8
SLC47A2
MATE 2 Fusion Protein Ag25472
20 μL
Primary
Canine, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers