Learn More
Masterflex™ Tygon™ E-LFL Pump Tubing
Description
- Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Regulatory approvals to match your unique application
Specifications
Specifications
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 1.42 mm |
| Material | Tygon E-LFL |
| Overall Length (Metric) | 3000 cm |
| For Use With (Application) | general laboratory use and provides longer life with peristaltic tubing pumps |
| Min. Operating Temperature (Metric) | -46°C |
| Packaging Quantity | 1 Each |
| Tubing Size | 34 |
| Max. Operating Temperature (Metric) | 74°C |
| Product Type | Pump Tubing |