Learn More
Masterflex™ Translucent C-Flex Braided Tubing
Product Code. p-8006898
Change view
Click to view available options
Diameter (Metric) Outer:
12.7 mm
19.05 mm
Unit Size:
25ft
Each
Product Code. Diameter (Metric) Outer unitSize
15436743 12.7 mm Each
15262700 19.05 mm 25ft
2 options
Product Code. 15436743
Brand: Masterflex™ 0643303
Only null left
Specifications
Certifications/Compliance FDA compliant, and complies with USP Class VI standards
Autoclavable Autoclavable
Diameter (Metric) Inner 6.4 mm
Material C-Flex
Diameter (Metric) Outer 12.7 mm
Overall Length (Metric) 7.62 m
For Use With (Application) For use with cell cultures, vessel and tank transfer, pump applications and cold storage.
Min. Operating Temperature (Metric) -45°C
Packaging Quantity 1 Pack
Color Translucent
Max. Operating Temperature (Metric) 135°C