Learn More
Masterflex™ L/S™ Easy-Load™ II Pump Head Mounting Hardware
Description
Mount up to four pump heads on a single drive. Masterflex™ Stainless Steel Mounting Hardware is designed for use with L/S™ Easy-Load™ II Pump Heads.
- For use with L/S Easy-Load II Pump Heads
- See individual drive specifications for maximum number of heads to be mounted
- Mounting hardware is stainless steel
Specifications
Specifications
| For Use With (Application) | Mounting L/S Easy-Load II pump head |
| For Use With | (1) L/S Easy-Load II pump heads |
| Quantity | 2 Pack |
| Type | Mounting Hardware |
| Case Material | Stainless Steel |