Learn More
Masterflex™ L/S™ Platinum-Cured Silicone Pump Tubing, spooled
Click to view available options
Tubing Size:
13
14
15
16
18
24
25
35
37
Unit Size:
400ft
Each
Description
Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
- Ensure that you always have enough tubing on hand
- Slightly greater clarity
- Preferred by pharmaceutical and biotechnology industries
- Fewer potential leachables
- USP Class VI
- FDA compliant
Specifications
Specifications
| Diameter (Metric) Inner | 6.4 mm |
| Material | Platinum-cured Silicone |
| For Use With (Application) | Pharmaceutical and biotechnology industries |
| Min. Operating Temperature (Metric) | -50°C |
| Packaging Quantity | 1 Ea. |
| Tubing Size | 24 |
| Flow Rate | 2.8 to 1700 mL/min. |
| Hose Barb | 1/4 in. |
| Maximum Pressure | 40 psi |
| Vacuum Level | 660 mm Hg |
| Show More |