Learn More
Sartorius™ Masterflex™ Economy Drive Peristaltic Pump
Product Code. 10728231 Shop All Sartorius Products
Change view
Click to view available options
Voltage:
230 V
Unit Size:
Each
Product Code. Voltage unitSize
10728231 230 V Each
1 options
Product Code. 10728231
Brand: Sartorius™ VFP001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Optimally suited to run Vivaflow™ laboratory crossflow cassette units. Sartorius™ Masterflex™ Economy Drive Peristaltic Pump is durable and cost-effective.
  • Suitable for Vivaflow™ laboratory crossflow cassette units
  • Durable and affordably priced
  • Variable speed
TRUSTED_SUSTAINABILITY
Specifications
Speed Control Variable
Weight (Metric) 4.1 kg
Voltage 230 V
Wattage 37 W