Learn More
Masterflex™ Norprene™ I/P™ High Performance Precision Pump Tubing
Product Code. 11721259
Click to view available options
Tubing Size:
70
88
89
Unit Size:
Each
Product Code. 11721259
Brand: Masterflex™ 0640488
Only null left
Add to basket
Add to basket
Outlast and outperform other tubing with 10,000 hours of tubing life
  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Always in stock — most formulations and sizes available to ship same day
  • Economical tubing with a longer tubing life
Specifications
Autoclavable Autoclavable
Diameter (Metric) Inner 12.7 mm
Material Norprene
Overall Length (Metric) 760 cm
For Use With (Application) applications requiring excellent chemical, heat, ozone and ultraviolet (UV) light resistance
Min. Operating Temperature (Metric) -51°C
Resistance Properties Acids
Tubing Size 88
Flow Rate 0.02 to 17L/min.
Hose Barb 1/2 in.
Color Opaque Black
Maximum Pressure 35 psi
Vacuum Level 660 mm
Max. Operating Temperature (Metric) 132°C
Product Type Pump Tubing