Learn More
Masterflex™ L/S™ Replacement Gear Assembly for 6 to 600 rpm Drives
Product Code. 11809721 Shop All Masterflex Products
Change view
Click to view available options
Speed:
6 to 600 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Unit Size
11809721 6 to 600 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11809721 Supplier Masterflex™ Supplier No. 0755309
  • ONLINE EXCLUSIVE PRICE
    1370.00 NOK
    1248.54 NOK / Pack of 1
    Save 121.46 NOK
    8% Off
Estimated Shipment: 07-01-2027
Make the most of your Masterflex™ drives replacement parts and service kits
  • Easy to install, these service kits avoid costly downtime of your gear pump head
  • Suitable for use with Masterflex™ L/S™ 6 to 600 rpm drives
Specifications
Type Replacement Parts
Speed 6 to 600 rpm