Learn More
Masterflex™ Mounting Kit
Easy stack multiple pump heads on your Masterflex™ L/S™ drive
Brand:  Masterflex™ 77800-00
Additional Details : Weight : 0.80000kg
Product Code. 11822603
  • 865.00 NOK / Each
Estimated Shipment: 21-05-2024
Description
  • Constructed of durable stainless steel
  • See drive specifications for recommended number of heads
  • Suitable for use with one Easy-Load™ 3 pump head
Specifications
Specifications
For L/S easy-load 3 pump head to Masterflex L/S pump drives
Single-channel mounting hardware and a 15 in. length of Silicone pump tubing
1 Each
Mounting Kit