Learn More
Masterflex™ L/S™ High-Performance Precision Pump Tubing, Tygon™ E-Food B-44-4X
Product Code. 15510652
Click to view available options
Tubing Size:
L/S 15
L/S 24
L/S 36
Unit Size:
15m
Each
Product Code. 15510652
Brand: Masterflex™ 0641836
Only null left
Add to basket
Add to basket
Masterflex™ L/S™ High-Performance Precision Pump Tubing is designed specifically for handling food products
  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
  • Applications: Food and beverage processing or dispensing
  • Characteristics: Nonwetting, odorless, tasteless, non-toxic, transparent
  • Specifications
    Autoclavable Autoclavable
    Diameter (English) Inner 0.382 in.
    Diameter (Metric) Inner 9.7 mm
    Material Tygon E-Food (B-44-4X)
    Overall Length (English) 50 ft.
    Overall Length (Metric) 15.2 m
    For Use With (Application) Food and beverage processing or dispensing
    Tubing Size L/S 36
    Flow Rate Max 3400 mL/min
    Hose Barb 3/8 in.
    Color Transparent
    Product Type Peristaltic Tubing