Learn More
Masterflex™ Tygon™ Clear E-Lab Pump Tubing
Product Code. p-8001551
Change view
Click to view available options
Tubing Size:
24
26
35
82
88
89
Unit Size:
15m
Each
Product Code. Tubing Size unitSize
13578990 24 15m
13509000 26 Each
13588990 35 Each
13539000 82 Each
13549000 88 Each
13559000 89 Each
6 options
Product Code. 13578990
Brand: Masterflex™ 0650924
Only null left
Add to basket
Add to basket
Use this tubing for general laboratory applications. Clear Tygon™ E-Lab Pump Tubing is ideal for inorganic chemicals and viscous fluids.
  • Clear tubing for easy flow monitoring
  • Nonaging, nonoxidizing
  • Low gas permeability
  • High dielectric constant
Specifications
Autoclavable Autoclavable
Diameter (Metric) Inner 6.4 mm
Material Tygon E-Lab
Overall Length (Metric) 1500 cm
For Use With (Application) general transfer applications
Min. Operating Temperature (Metric) -46°C
Packaging Quantity 1 Each
Tubing Size 24
Flow Rate 2.8 to 1700mL/min.
Hose Barb 1/4in.
Height (Metric) 8.8 m
Color Clear
Maximum Pressure 40 psi
Vacuum Level 660 mm
Max. Operating Temperature (Metric) 74°C
Product Type Pump Tubing