Learn More
Eppendorf™ Mastercycler Nexus Flat Eco, EU-Plug
Versatile, quiet, and energy efficient PCR cycler.
Brand:  Eppendorf™ 6330000013
Additional Details : Weight : 10.50000kg
Product Code. 15755675
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Intuitive graphic programming
  • Small footprint
  • Universal block for ultimate flexibility
  • Connect up to 2 Mastercycler nexus eco to your existing unit
  • Available with gradient
  • E-mail notification
  • flexlid™ concept allows use of all types of consumables with automatic height adjustment of the lid
Specifications
Specifications
> 700
Lid: 37-110°C, Block: 4-99°C
230 V, 50/60 Hz
CAN_in/CAN_out, Ethernet, USB
10.4 kg
EU Plug
25 × 41.2 × 32.1 cm
4 slides or other matching consumables
In situ PCR